1x1 image pixel for data collection
Enviro-Safe Thermometer
Enviro-Safe Thermometer
Actual product may vary
Fisherbrand
Enviro-Safe Thermometer
Use in place of hazardous spirit or mercury filled thermometers. Bel Art™ Enviro-Safe™ thermometers are environmentally safe and filled with a biodegradable liquid that can be cleaned up with soap and water.
Have Questions?
Catalog NumberTemperature Range (Metric)
FKS11737093-20°C to +150°C
Catalog number FKS11737093
Price (SGD)
-
Temperature Range (Metric):
-20°C to +150°C
Enviro-Safe™ thermometers are environmentally safe and filled with a biodegradable liquid that can be cleaned up with soap and water. They can be used in place of hazardous spirit or mercury filled thermometers. The glass is lead free and non-toxic and the packaging is recyclable.
  • Non-toxic
  • Non-hazardous
  • Environmentally friendly
  • Lead free glass
  • Accuracy traceable to NIST and DKD (statement of accuracy included)
Specifications
Certifications/ComplianceNIST, DKD
Immersion TypePartial
Length (Metric)305 mm
Product TypeThermometer
Max. Temperature (Metric)150°C
Min. Temperature (Metric)-20°C
Temperature Range (Metric)-20°C to +150°C
CalibratedYes
Accuracy±1°C
Immersion Length76 mm
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link