Stainless Steel Tweezers
Stainless Steel Tweezers
Actual product may vary
Fisherbrand
Stainless Steel Tweezers
Recommended for micro electronics and precision assembly
Have Questions?
Catalog NumberLength (Metric)Tip Style
FKS1173544910.8 cmUltra Fine Sharp
Catalog number FKS11735449
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
10.8 cm
Tip Style:
Ultra Fine Sharp
Recommended for micro electronics and precision assembly
  • Hand finished points for accuracy and excellent tip hardness
  • Anti-glare finish reduces eye strain
  • Anti-magnetic and acid resistant
Specifications
Tip StyleUltra Fine Sharp
No. per PackEach
Length (English)4.25 in.
MaterialStainless Steel
Length (Metric)10.8 cm
Product TypeTweezers
TypeTweezer
QuantityEa.
StyleStraight
Unit SizeEA