Check Order Status
Angled Pipette Holder
Actual product may vary
Fisherbrand
Angled Pipette Holder
This Angled Pipette Holder provides safe, convenient storage and easy retrievel of your pipettes.
Have Questions?
Catalog number
FKS11734949
Price (SGD)
1,151.00
EA
Add to cart
Price (SGD)
1,151.00
EA
Add to cart
Keeps pipettes handy and well-organized.
Specifications
Product Type
Pipet Rack
Quantity
1 Ea.
Width (Metric)
88.9 mm
Width (English)
3.5 in.
Height (Metric)
406.4 mm
Height (English)
16 in.
Depth (English)
11.5 in.
Depth (Metric)
292.1 mm