Gosselin™ Polystyrene 10mL Round-Base Tubes
Gosselin™ Polystyrene 10mL Round-Base Tubes
Actual product may vary
Fisher Catalogue
Gosselin™ Polystyrene 10mL Round-Base Tubes
Manufactured from polystyrene. Gosselin™ Polystyrene 10mL Round-Base Tubes are appropriate for clinical, food, and industrial settings.
Have Questions?
Catalog NumberDiameter (Metric)Closure Type
FKS1172939215.4 mmStraight Cap
Catalog number FKS11729392
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
15.4 mm
Closure Type:
Straight Cap
  • Round base
  • Polystyrene
  • Natural (clear) color
  • Safe for food
  • Certified EC N°1935/2004, ISO 11137 : Nil
Specifications
Certifications/ComplianceISO 11137
Closure ColorRed
Closure MaterialLDPE
ColorNatural
Capacity (Metric)10 mL
MaterialPolystyrene
DisposableReusable
For Use WithFor clinical, dairy, environment, industries, microbiology
Packaging250BG
Quantity1000 Carton
RimmedNo
SterilityNon-sterile
Diameter (Metric)15.4 mm
Closure TypeStraight Cap
Product TypeTest Tube
Unit SizePK