Classic Vortex Mixer
Classic Vortex Mixer
Actual product may vary
Fisherbrand
Classic Vortex Mixer
Combines the highest performance ratings in terms of speedwith excellent reliability and safety. Fisherbrand™ Classic Vortex Mixer includes two operating modes, the possibility to change the vibration frequency, and a wide range of accessories.
Have Questions?
Catalog number FKS11726744
Price (SGD)
1,021.00
EA
Add to cart
Price (SGD)
1,021.00
EA
Add to cart
Combines the highest performance ratings in terms of speedwith excellent reliability and safety. Fisherbrand™ Classic Vortex Mixer includes two operating modes, the possibility to change the vibration frequency, and a wide range of accessories.
Original Design
  • Compact in height and area
  • Stability is optimized with selection lever related to operating mode
  • 3 anti-slip rubber feet for staying in required position
Multi-Environment Operation
  • Variable speed operation, electronically controlled up to 3000 rpm
  • Large choice of accessories to suit tubes, plates and flasks
  • Universal voltage / frequency model
Designed to Last
  • Accessories fit by simple pressure
  • Optimum resistance to chemicals from technopolymer casing
  • Excellent stability with zinc alloy base
  • IP 42 protection against particles and liquids
Specifications
Orbit4.5 mm
Speed Range0 to 3000 rpm
Wattage15 W
Product TypeVortex Mixer
Frequency50/60 Hz
MaterialZinc Alloy Base, Technopolymer Platform
Depth (Metric)220 mm
Dimensions (L x W x H)7.1 x 8.7 x 2.8 in. (180 x 220 x 70 mm)
Height (English)2.75 in.
Height (Metric)70 mm
Length (English)7.1 in.
Length (Metric)180 mm
Width (English)8.7 in.
Width (Metric)220 mm
Weight (Metric)2.7 kg
ModelClassic
IP RatingIP 42
Operating ModesTouch, Continuous
Unit SizeEA