SP Scienceware™ PTFE Magnetic Stirring Bar Kit
SP Scienceware™ PTFE Magnetic Stirring Bar Kit
Actual product may vary
Fisherbrand
SP Scienceware™ PTFE Magnetic Stirring Bar Kit
Supplied in a convenient storage box and provide instant availability of PTFE encapsulated Alnico magnets.
Have Questions?
Catalog NumberQuantityShape
FKS117250643 sizes (2 each size)Octagon
Catalog number FKS11725064
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
3 sizes (2 each size)
Shape:
Octagon
  • Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
  • Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
  • Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification
  • Autoclave safe
Specifications
AutoclavableAutoclavable
Certifications/ComplianceISO 9001:2015
ColorWhite
Dimensions (L x W x H)25.4 x 8mm, 38.1 x 8mm, 50.8 x 8mm (Two Each)
For Use With (Equipment)Vials, Micro test tubes, Centrifuge tubes
MaterialAlnico Magnet, PTFE
Quantity3 sizes (2 each size)
ShapeOctagon
Unit SizePK