DURAN™ High Temperature red EFTE Pouring ring, GL 32
DURAN™ High Temperature red EFTE Pouring ring, GL 32
Actual product may vary
Fisherbrand
DURAN™ High Temperature red EFTE Pouring ring, GL 32
Screw cap with cap liner sealing and high temperature and chemical resistance.
Have Questions?
Catalog NumberThread Size (Metric)
FKS11722342GL 32
Catalog number FKS11722342
Price (SGD)
67.00
PK
Add to cart
Thread Size (Metric):
GL 32
Price (SGD)
67.00
PK
Add to cart
The red polybutylene terephthalate (PBT) GL cap and ETFE pouring rings offer greater thermal and chemical resistance than the equivalent polypropylene components. Screw cap seal has a PTFE faced cap liner for efficient sealing. ETFE pouring ring ensures drip free pouring.
  • replacement, cap, liner, silicone, seal, sealing, vmq, ptfe, coated, screw, cap
  • Excellent chemical resistance due to PTFE coating
  • Cap liner forms a tight seal with bottle Peroxide catalysed silicone
  • Peroxide catalysed silicone
  • Suitable for PBT screw caps and tamper-evident screw caps
  • Fully compliant with current international food contact regulations
Specifications
ColorRed
DescriptionPouring ring
For Use With (Application)Autoclaving of media and reagents; Preparation of media and reagents; Suitable for dry heat sterilization; Pressure applications; Storage of corrosive reagents; Chemical and thermal reactions; Long term / archival storage of samples, media and reagents; Sampling; Transport and shipping
MaterialPBT, ETFE, Fiberglass
Quantity10/Pk.
Screw MaterialETFE
Thread Size (Metric)GL 32
Liner MaterialPTFE-coated Silicone
AutoclavableAutoclavable
Closure MaterialETFE
Unit SizePK