Wheaton™ Arsine Generator, Modified
Wheaton™ Arsine Generator, Modified
Actual product may vary
Fisherbrand
Wheaton™ Arsine Generator, Modified
Modified for Arsine analysis
Have Questions?
Catalog number FKS11722235
Price (SGD)
699.00
EA
Add to cart
Price (SGD)
699.00
EA
Add to cart
  • Modified version of catalog number 180020 for arsine analysis
  • The freestanding system has a standard taper 24/40 Clear-Seal™ joint
  • One piece scrubber/absorber unit
  • Low extractable borosilicate glass
  • Eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes
  • Manufactured from WHEATON 33 low-extractable borosilicate glass
Specifications
Product TypeArsine Generator
Standard Taper Joint24/40
MaterialBorosilicate Glass
Certifications/ComplianceUSP Type I and ASTM E438 Type I, Class A
For Use With (Application)For arsine analysis using SDDC method
Includes125mL erlenmeyer flask with 24/40 joint, one-piece scrubber/absorber unit
Unit SizeEA