Secador™ Auto-Desiccator Module Desiccant Replacement Beads, 26 g
Secador™ Auto-Desiccator Module Desiccant Replacement Beads, 26 g
Actual product may vary
Secador™ Auto-Desiccator Module Desiccant Replacement Beads, 26 g
Secador™ Auto-Desiccator Module Desiccant Replacement Beads, 26 g
Secador™ Auto-Desiccator Module Desiccant Replacement Beads, 26 g
Fisherbrand
Secador™ Auto-Desiccator Module Desiccant Replacement Beads, 26 g
Color-indicating silica gel beads for use in refilling the Secador auto-desiccator module.
Have Questions?
Catalog number FKS11721436
Price (SGD)
108.00
EA
Add to cart
Price (SGD)
108.00
EA
Add to cart
Color-indicating silica gel beads for use in refilling the Secador auto-desiccator module
  • Automatically regenerates for thousands of cycles.
  • Absorbs moisture from the air.
  • Inert, non-toxic, non-corrosive.
  • Will not disintegrate into powder even after repeated regeneration.
  • Color-changing moisture indicator turns from blue to light pink indicating saturation.
  • Pink/dark brown hues indicate replacement due.
Specifications
Diameter (Metric) Outer5 cm
Product TypeDesiccant Replacement Bead
MaterialSilica Gel
For Use With (Application)Color-indicating silica gel beads for use in refilling the Secador™ auto-desiccator module
Weight (Metric)26 g
Unit SizeEA