Check Order Status
Grade 501 Chromatography Paper
Actual product may vary
Fisherbrand
Grade 501 Chromatography Paper
Fisherbrand™ Grade 501 Chromatography Paper is standard grade chromatography paper for routine or simple analysis.
Have Questions?
Catalog Number
Length (Metric)
FKS11718742
200 mm
Catalog number
FKS11718742
Price (SGD)
167.00
PK
Add to cart
Length (Metric):
200 mm
Price (SGD)
167.00
PK
Add to cart
Grade 501
Standard chromatography paper for routine and simple analysis
Specifications
Film Thickness
0.19 mm
For Use With (Application)
Standard Chromatography Paper for Routine and Simple Analysis
Format
Sheet
Grade
501
Material
Paper
Width (Metric)
200 mm
Flow Rate
145 mm/30 min.
Length (Metric)
200 mm