Difco™ Dehydrated Culture Media: Marine Agar 2216
Difco™ Dehydrated Culture Media: Marine Agar 2216
Actual product may vary
Fisherbrand
Difco™ Dehydrated Culture Media: Marine Agar 2216
Isolation, cultivation and enumeration of heterotrophic marine bacteria. BD Difco™ Dehydrated Culture Media: Marine Agar 2216 is part of a large selection of dehydrated media.
Have Questions?
Catalog number FKS11718183
Price (SGD)
2,243.00
EA
Add to cart
Price (SGD)
2,243.00
EA
Add to cart
  • Dehydrated
  • Culture Media
  • Marine Agar 2216
Specifications
FormDehydrated
Media TypeMarine Agar 2216
Product TypeAgar
FormulationPeptone 5g/L, Yeast extract 1g/L, Ferric Citrate 0.1g/L, Sodium Chloride 19.45g/L, Magnesium Chloride 8.8g/L, Sodium Sulfate 3.24g/L, Calcium Chloride 1.8g/L, Potassium Chloride 0.55g/L, Sodium Bicarbonate 0.16g/L, Potassium Bromide 0.08g/L, Strontium Chloride 34mg/L, Boric Acid 22mg/L, Sodium Silicate 4mg/L, Sodium Fluoride 2.4mg/L, Ammonium Nitrate 1.6mg/L, Disodium Phosphate 8mg/L, Agar 15g/L
For Use With (Application)Isolation, cultivation and enumeration of heterotrophic marine bacteria
Packaging TypeBottle
Quantity500 g
Shelf Life1095 Days
Unit SizeEA
Contents & Storage
<30°C