1x1 image pixel for data collection
DURAN™ Laboratory Bottle, Square, Clear, with DIN 168-1 Thread, Graduated
DURAN™ Laboratory Bottle, Square, Clear, with DIN 168-1 Thread, Graduated
Actual product may vary
Fisherbrand
DURAN™ Laboratory Bottle, Square, Clear, with DIN 168-1 Thread, Graduated
Multifunctional glass laboratory bottles with square space saving shape.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11716233100 mL
FKS11321465250 mL
FKS11331465500 mL
FKS113414651000 mL
Catalog number FKS11716233
Price (SGD)
365.00
PK
Add to cart
Capacity (Metric):
100 mL
Price (SGD)
365.00
PK
Add to cart
DURAN™ Square laboratory bottles have a space saving shape that offers the benefit of close and efficient packing. With up to a 20 % saving of space compared to traditional round bottles. Ideal for the shipping and storage of high-demand materials. For reagent supply to instruments where bench space is at a premium. The bottles are moulded from high-purity, clear, USP/EP Type I DURAN™ borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ square bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140°C. All components are fully autoclavable at 121 or 134°C.
  • Ergonomic handling, high stability and good stackability due to angular shape
  • Saves space and costs in autoclaves, drying chambers, ovens, storage or washers
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
  • Easy-to-read scale with graduation marks to estimate the content
  • Fired-on, highly durable white ceramic
Specifications
Capacity (Metric)100 mL
For Use With (Application)Liquid handling reservoir for instruments; Transport and shipping; Sample collection; Storage of media and reagents; Archival storage of samples; Preparation of media and reagents; Culturing vessel
IncludesWith screw cap and pouring ring from PP (blue)
MaterialBorosilicate Glass (3.3)
Quantity10/Pk.
Thread Size (Metric)GL 32
AutoclavableAutoclavable
Height (Metric)113 mm
Closure MaterialPolypropylene
Diameter (Metric)50 mm
Unit SizePK
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link