Digital Thermometers with Stainless-Steel Probe on Cable
Digital Thermometers with Stainless-Steel Probe on Cable
Actual product may vary
Fisher Catalogue
Digital Thermometers with Stainless-Steel Probe on Cable
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)Temperature Range (English)AccuracyLength (Metric) Cable
FKS11715843-50°C to +300°C-58°F to +572°F±1°C305 cm
CTL15-077-9E-50°C to +300°C-58°F to +572°F±1°C305 cm
CTL15-077-32-50°C to +150°C-58°F to +302°F±1°C between -30 to 150°C, otherwise ±2°C45.72 cm
CTL14-648-46-50°C to +300°C-58°F to +572°F±1°C from -20°C to +100°C73.7 cm
CTL15-077-9DN/A-58°F to +572°F±1°C304.8 cm
CTL15-077-290°C to 200°C32°F to 392°F±2°C106.6 cm
Catalog number FKS11715843
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
-50°C to +300°C
Temperature Range (English):
-58°F to +572°F
Accuracy:
±1°C
Length (Metric) Cable:
305 cm
Easy-to-read digital thermometers are compact to hold in your hand or mount on wall in restricted areas. Fisher Scientific™ Digital thermometer with stainless-steel probe is chemically resistant to most acids, bases and solvents and may be used to measure temperature of gases, liquids, food, and other semisolids.
  • Models 15-077-16, -15, -21, -8B, -29 and 14-648-46 have alarm that can be set to alert user to temperatures that exceed setpoints
  • Some models (such as 15-077-16 and -15) have special features that print temperature, high/low memory and time of day; 15-077-29 thermometer has additional timer alarms
  • For a miniature-size thermometer, see model 15-077-32
For use in cuvets, soil, incubators, petroleum products and piercing food samples. Waterproof thermometers (models 15-077-9E, -9D and 14-648-46) are ideal for wet labs, wash-down areas and outdoor applications in the agricultural, petrochemical, food, and pharmaceutical industries.
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Diameter (English) Probe0.14 in.
For Use With (Application)Wet labs, wash down areas and outdoor applications in the agricultural, petrochemical, food and pharmaceutical industries
IncludesSuction cup, magnets, Velcro™, traceable certificate and 1.5V Silver Oxide battery
Length (English) Cable120 in.
Length (English) Probe5 in.
Length (Metric)121 mm
Length (Metric) Cable305 cm
Length (Metric) Probe12.7 cm
MaterialStainless Steel
Resolution0.1°C
Weight (English)2 oz.
Weight (Metric)56.6 g
Product TypeTraceable Digital Thermometer w/Probe
Length (English)4.75 in.
Max. Temperature (English)572°F
Max. Temperature (Metric)300°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +572°F
Temperature Range (Metric)-50°C to +300°C
CalibratedYes
Accuracy±1°C
DisplayDigital LCD
Width (Metric)8 cm
Unit SizeEA