Store and transport 20 or 30 standard microscope slides in sturdy cardboard folders. Cardboard Slide Trays feature recessed slots that protect specimens and thumb cuts for easy slide access. Disposable trays are ideal for short-term storage or slide transporting.
Have Questions?
Change view
Catalog Number
Color
Slide Capacity
FKS11713217
Black
30
FIS12-587-10
Black
20
FKS11703217
Black
20
3 Options
Catalog number FKS11713217
Price (SGD)
33.00
PK
Add to cart
Color:
Black
Slide Capacity:
30
Price (SGD)
33.00
PK
Add to cart
Store and transport up to 20 standard microscope slides in sturdy cardboard folders. Fisherbrand™ Cardboard Slide Trays feature recessed slots that protect specimens and thumb cuts for easy slide access. Disposable trays are ideal for short-term storage or slide transporting.
Accommodates standard microscope slides (25 x 75 mm and 1 x 3 in.)
Horizontal compartments are recessed to protect the specimens when the cover is closed
Features thumb cuts for easy slide removal
Folder cover includes a number slide identification sheet
Disposable folders are available in 2 sizes and different colors
Specifications
Slide Capacity30
ColorBlack
Compartments30
Device Holds30 Slides
Dimensions (L x W x H)281 x 328 x 10 mm
For Use With (Application)For storage or transport
For Use With (Equipment)Standard Microscope Slides (25 x 75 mm)