AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Dimensions (L x W x H)195 x 95mm
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Height (Metric)30mm
Includes12 Oval Stirrer Bars: 2 each of 10 x 5 mm, 15 x 6 mm, 25 x 10 mm, 30 x 10 mm, 35 x 13 mm; 1 each of 50 x 17 mm, 70 x 27 mm
Width (Metric)95mm
CoatingPTFE
Length (Metric)195 mm
Quantity1/Ea.
TypePTFE Stirrer Bars Box Set2
Unit SizeEach