Check Order Status
Carbon Steel Scalpel Blades
Actual product may vary
Fisherbrand
Carbon Steel Scalpel Blades
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
Have Questions?
Change view
Catalog Number
Size
Length (Metric)
FKS11702734
# 23
30 mm
FKS11748373
# 11
20 mm
2 Options
Catalog number
FKS11702734
Price (SGD)
70.00
PK
Add to cart
Size:
# 23
Length (Metric):
30 mm
Price (SGD)
70.00
PK
Add to cart
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
Carbon steel to BS EN 27740
Handles and blades sold separately
Specifications
Blade Material
Carbon Steel
Product Type
Scalpel Blade
Size
# 23
Length (Metric)
30 mm
For Use With (Equipment)
The # 23 fits Handles # 4, # 4L, # 4 Graduated and # 6B
Disposable
Single-use
Certifications/Compliance
BS EN 7740
Quantity
10/Pk.