Star-24 LED Stereo Microscope
Star-24 LED Stereo Microscope
Actual product may vary
Fisher Catalogue
Star-24 LED Stereo Microscope
Particularly well suited to viewing specimens at low magnifications and introduces the benefits of LED lighting into the stereo range of microscopes.
Have Questions?
Catalog number FKS11697630
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Designed for both educational and professional use
  • Diamond-shaped base with sloping sides provides an ergonomic hand rest during sample manipulation to ensure control and minimize the possibility of damage occurring
  • Binocular optical head, inclined at 45°
  • Interpupillary distance adjustment (Siedentopf type) 50 to 75mm
Specifications
Magnification Power2X, 4X
Zoom Range40mm
Electrical Requirements220/240 V 50/60 Hz
Head StyleBinocular, 45° Inclined
Lighting TypeLED, Variable
EyepieceWF 10X/20 mm
Unit SizePK