Check Order Status
Maxi Pipet Tips
Actual product may vary
Fisherbrand
Maxi Pipet Tips
For use with large volume pipets
Have Questions?
Change view
Catalog Number
Volume (Metric)
FKS11648138
5000 μL
FIS21-195-3
1000 to 5000 μL
FIS02-707-467
1 to 5 mL
FIS02-681-410
5000 μL
4 Options
Catalog number
FKS11648138
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
5000 μL
For use with large volume pipets
Designed for use when transferring large sample volume
Advanced molding technology ensure tips are straight
Alternative option to serological pipets
Nonsterile
Certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free
Specifications
Volume (Metric)
5000 μL
Sterility
Non-sterile
Color
Natural
Autoclavable
Autoclavable
Length (English)
4.75 in.
Length (Metric)
150 mm
Format
Bulk
Tip Style
Universal, Macro
For Use With (Equipment)
Eppendorf Research, Jencons Sealpette, Genemate ISC Bio Express
Graduated
No
Material
Polypropylene