Check Order Status
Natural Rubber One Hole Stoppers
Actual product may vary
Fisher Catalogue
Natural Rubber One Hole Stoppers
Used for laboratory purposes.
Have Questions?
Change view
Catalog Number
Diameter (Metric) Bottom
Diameter (Metric) Top
FKS11597612
62 mm
69 mm
FKS11527602
19 mm
22.5 mm
FKS11577602
29 mm
33 mm
FKS11587612
57 mm
64 mm
FKS11507622
67 mm
74 mm
5 Options
Catalog number
FKS11597612
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric) Bottom:
62 mm
Diameter (Metric) Top:
69 mm
Made up of natural rubber
Red in color
Contains one hole
Specifications
Diameter (Metric) Hole
5mm
For Use With (Application)
For laboratory use
Hardness
46/55 IRHD
Length (Metric)
49 mm
Quantity
2/Pk.
Color
Red
Diameter (Metric) Bottom
62 mm
Diameter (Metric) Top
69 mm
Material
Natural Rubber
Product Type
Tapered Stopper (1 hole)