Search Thermo Fisher Scientific
Catalog Number | Diameter (Metric) Outer | Diameter (Metric) Inner | Color |
---|---|---|---|
FKS11592493 | 41 mm | 19 mm | Red |
FKS11542493 | 21 mm | 11 mm | Red |
FKS11552493 | 25 mm | 12.5 mm | Red |
FKS11532503 | 57 mm | 25 mm | Red |
FKS11522503 | 53 mm | 24 mm | Red |
FISFB68685 | 29 mm | 14 mm | Cream |
FISFB68684 | 25 mm | 12.5 mm | Cream |