Grade 912 Universal Test Paper
Grade 912 Universal Test Paper
Actual product may vary
Fisherbrand
Grade 912 Universal Test Paper
Used for general analysis.
Have Questions?
Catalog NumberTypeColor IndicatorDimensions (L x W)Format
FKS11592413KI Starched PaperWhite60 x 10 mmTube
Catalog number FKS11592413
Price (SGD)
-
Type:
KI Starched Paper
Color Indicator:
White
Dimensions (L x W):
60 x 10 mm
Format:
Tube
  • General control of acid or alkaline reactions
Color:
  • Litmus Blue: Blue to red
  • Litmus neutral: Purple to blue/red
  • Litmus red: Red to blue
Applications:
  • Rapid determination of pH value in industrial applications
  • General analysis in school
  • Used in formation
Specifications
Color IndicatorWhite
FormatTube
For Use With (Application)Rapid determination of pH value in industrial applications, general analysis in school, formation
Quantity200 Pk.
TypeKI Starched Paper
Dimensions (L x W)60 x 10 mm
Length (Metric)60 mm
Width (Metric)10 mm
Unit SizeEA