Stainless Steel Blunt End Forceps
Stainless Steel Blunt End Forceps
Actual product may vary
Fisherbrand
Stainless Steel Blunt End Forceps
Provides a non-stick surface with PTFE coating to which very few solid or liquid substances will permanently adhere. Nickel Electro™ Stainless Steel Blunt End Forceps with excellent non-wetting properties provide easy cleaning and can be steam or chemically sterilized.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric) Body
FKS11592113115 mm
FKS11502123130 mm
Catalog number FKS11592113
Price (SGD)
66.00
EA
Add to cart
Height (Metric) Body:
115 mm
Price (SGD)
66.00
EA
Add to cart
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.
  • Chemically resistant and non-contaminating
  • Non-protein binding and providing bio-inert surfaces
  • No critical quantities lost due to sticking
  • No build up on surfaces
  • Ideal for use under severe laboratory conditions
  • Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized
  • With guide pin
Specifications
Tip StyleBlunt
No. per PackEach
MaterialPTFE-coated Stainless Steel
Product TypeForceps
Height (Metric) Body115 mm
StyleSerrated Jaws
Unit SizeEA