Check Order Status
Borosilicate Glass Graduated Cylinder
Actual product may vary
Fisherbrand
Borosilicate Glass Graduated Cylinder
Measure liquids with blue ceramic scale graduated glass cylindars. The Fisherbrand™ borosilicate glass graduated cylindawith hexagonal base and spout
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Graduations
FKS11587702
2000 mL
20 mL
FKS11517702
10 mL
0.2 mL
FKS11527702
25 mL
0.5 mL
FKS11567702
500 mL
5 mL
4 Options
Catalog number
FKS11587702
Price (SGD)
-
Capacity (Metric):
2000 mL
Graduations:
20 mL
DIN 12680, BS 604, ISO 4788
Graduated in blue ceramic
Borosilicate Glass
Hexagonal base
Spouted
Individual reorder code on each product
Specifications
Base Type
Hexagonal
Scale Color
Blue
Type
Graduated Cylinder
Material
Borosilicate Glass
Certifications/Compliance
BS 604, DIN 12680, ISO 4788
Class
B
Capacity (Metric)
2000 mL
Scale
Blue Ceramic
Graduations
20 mL