Borosillicate Glass Funnel
Borosillicate Glass Funnel
Actual product may vary
Fisher Catalogue
Borosillicate Glass Funnel
Plain glass funnel manufactured from borosilicate glass. Fisherbrand™ Borosilicate Glass Funnel is ideal for use with funneling liquids in laboratory applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)TopDiameter (Metric) StemLength (Metric) Stem
FKS1158242375mm9mm80mm
FKS1157242355mm8mm60mm
Catalog number FKS11582423
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric)Top:
75mm
Diameter (Metric) Stem:
9mm
Length (Metric) Stem:
80mm
Plain glass funnel manufactured from borosilicate glass.
Specifications
Diameter (Metric)Top75mm
Diameter (Metric) Stem9mm
MaterialBorosilicate Glass
Length (Metric) Stem80mm
Unit SizePK