Provides a non-stick surface with PTFE coating to which very few solid or liquid substances will permanently adhere. Nickel Electro™ Stainless Steel Sharp Ends Forceps provide excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized.
Have Questions?
Change view
Catalog Number
Height (Metric) Body
FKS11582113
160 mm
FKS11562113
115 mm
2 Options
Catalog number FKS11582113
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Height (Metric) Body:
160 mm
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.
Chemically resistant and non-contaminating
Non-protein binding and providing bio-inert surfaces
No critical quantities lost due to sticking
No build up on surfaces
Ideal for use under severe laboratory conditions
Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized