Ideal for hundreds of lab applications, including counting blood samples, taking surveys, laboratory inventory, counting total drops of liquid, receiving lab samples and tallying biomedical events.
Have Questions?
Catalog number FKS11577483
Price (SGD)
-
Easy-to-use, four-digit unit records up to 9999
Numbers instantly and automatically return to zero by twisting the reset knob
Large white numerals appear on a black background
Protective window seals out dust, dirt and humidity
Rugged, heavy-duty chrome-finished case has a hinged finger loop for a sure grip
Finger pressure on the button produces an audible click that signals the count
Unique engineering provides a smooth, tactile feel with every count
Dimensions: 1.75 dia. × 1.13 in.W (4.4 × 2.9cm)
Weight: 3 oz.
Specifications
Product TypeHand Tally Counter
Displayed Text9999
QuantityEach
Diameter (English)1.75 in.
Diameter (Metric)4.45cm
Width (English)1.13 in.
Width (Metric)2.9cm
For Use With (Application)Ideal for lab applications