Made from chemically resistant virgin plastic, Bacti-Caps™ provide a firm, positive culture tube closure allowing a controlled gas interchange. They are ideal for use in science, research, and other laboratories.
Have Questions?
Catalog Number
Color
FKS11577143
White
Catalog number FKS11577143
Price (SGD)
138.00
PK
Add to cart
Color:
White
Price (SGD)
138.00
PK
Add to cart
Bacti-Caps™ for test tubes without edge
Provide a firm, positive culture tube closure allowing a controlled gas interchange, thus proving a more practical alternative to cotton wool plugs
Suitable for all standard test tube sizes, they fit rimless and plastic tubes with an outside diameter of 13mm, 16mm, 19mm, 25mm and 38mm
The three flexible fins molded on the inner wall of each cap ensure a positive fit even when minor variations in tube size occur
Bacti-Caps™ are made from chemically resistant virgin plastic so they are unaffected by culture media, most acids, alkalis, alcohols and esters