Translucent Bottles, MDPE
Translucent Bottles, MDPE
Actual product may vary
Fisher Catalogue
Translucent Bottles, MDPE
Use these general purpose bottles for liquid storage and sampling. Fisherbrand™ Translucent Bottles are leakproof and offer excellent chemical resistance. Autoclavable with cap loosened Ideal for general laboratory applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11573242500 mL
FKS11543242125 mL
Catalog number FKS11573242
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
500 mL
  • Clear general purpose bottles
  • Polypropylene screw closure
Specifications
Capacity (Metric)500 mL
ColorTranslucent
For Use With (Application)Ideal for general laboratory applications
IncludesPolypropylene screw closure
MaterialMDPE
No. per Pack9
Width (Metric)205mm
Closure IncludedYes
AutoclavableAutoclavable
MouthNarrow
Height (Metric)140 mm
Product TypeNarrow Neck Bottle
Closure MaterialPolypropylene
Closure Size58/R3
Diameter (Metric)76 mm
Closure TypeScrew Cap
Unit SizePK