Twirl'EM™ Clear Polyethylene Sampling Bags
Twirl'EM™ Clear Polyethylene Sampling Bags
Actual product may vary
Fisher Catalogue
Twirl'EM™ Clear Polyethylene Sampling Bags
Collect solid, semi-solid or liquid samples in the laboratory using Twirl'EM™ Clear Polyethylene Sampling Bags. The bags feature a twirl round metal wire closure system with safety tabs to provide a leakproof seal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS115727731240 mL
FKS11542773650 mL
Catalog number FKS11572773
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1240 mL
  • Sterile
  • Clear polyethylene
  • Without side seals, opening at width
  • Pull tabs to open bag without touching edges
  • Twirl round metal wire closure system with safety tabs
Specifications
Capacity (Metric)1240 mL
ColorClear
MaterialPolyethylene
SterilitySterile
Quantity1000 Pk.
Unit SizeEA