Lab Thermometer
Actual product may vary
Fisherbrand
Lab Thermometer
A general purpose/stirring thermometer for a wide application of laboratory needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)SubdivisionImmersion TypeLength (Metric)
FKS11571993-10°C to +150°C1°CPartial Immersion305 mm
FKS11587273-20°C to +110°C1°CTotal Immersion305 mm
FKS11597273-20°C to +150°C1°CTotal Immersion305 mm
ASO6-302-11-10°C to +50°C1°CTotal Immersion155 mm
FKS11517283-10°C to +110°C1°CTotal Immersion155 mm
FKS11551993-10°C to +50°C0.5°CPartial Immersion305 mm
FKS11561993-10°C to +110°C1°CPartial Immersion305 mm
Catalog number FKS11571993
Price (SGD)
12.00
Each
Temperature Range (Metric):
-10°C to +150°C
Subdivision:
1°C
Immersion Type:
Partial Immersion
Length (Metric):
305 mm
Price (SGD)
12.00
Each
  • With amber graduations
  • Non-roll fitting
  • Reinforced bulb for stirring
Specifications
For Use With (Application)General use
Immersion TypePartial Immersion
Length (Metric)305 mm
Thermometer FillRed Spirit
Product TypeLiquid Filled Thermometer
Max. Temperature (Metric)150°C
Min. Temperature (Metric)-10°C
Temperature Range (Metric)-10°C to +150°C
Subdivision1°C
Immersion Length76 mm
Unit SizeEach