Eutech™ CONSEN9501D Conductivity /Temperature Electrode
Eutech™ CONSEN9501D Conductivity /Temperature Electrode
Actual product may vary
Fisherbrand
Eutech™ CONSEN9501D Conductivity /Temperature Electrode
Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Have Questions?
Catalog NumberCell Constant
FKS11543845K = 1.0
Catalog number FKS11543845
Price (SGD)
889.00
EA
Add to cart
Cell Constant:
K = 1.0
Price (SGD)
889.00
EA
Add to cart
Eutech™ Conductivity Electrodes are used in conductivity meters in various applications. Get quick and accurate conductivity measurements in the field, laboratories, and schools.
Specifications
  • Parameter: Conductivity / Temperature
  • Range: 0 to 150mS
  • Temperature Range: 0 to 80 °C*
  • Cell: 2
  • Cell Constant: K = 1.0
  • Cellmaterial: Stainless steel rings, PP and ultem-body
  • ATC:Yes
  • Dimensions (Shaft) :44 x 16mm
  • Cable Length:1m
  • Connector:8-pin DIN
  • Used With: CON 700 / PC 700 / CON 2700 / PC 2700
Specifications
Cell ConstantK = 1.0
TypeConductivity/Temperature Electrode
MaterialUltem-Body
Measurement Ranges0 to 150 mS
Unit SizeEA