Swabs in Peel Packs
Swabs in Peel Packs
Actual product may vary
Fisherbrand
Swabs in Peel Packs
Use these swabs for taking samples. Fisherbrand™ Swabs have a round, compact and shred-resistant head.
Have Questions?
Catalog NumberdimTipType
FKS11542483
Catalog number FKS11542483
Price (SGD)
-
  • Swab with wooden shaft and cotton tip
  • Sterile by ethylene oxide
  • Dry in peel packs
  • 150mm x 5mm
  • Produced under strict ISO 9002/EN 29002 standards
  • CE marked No.98 06 0119, Class II A
Specifications
SterilitySterile
Certifications/ComplianceISO 9002/EN 29002, CE Marked No. 98 06 0119
Handle MaterialWood
Tip MaterialCotton
IncludesAll Swabs Have a Round, Compact and Shred-Resistant Head for Sample Taking
PackagingPeel Pack
For Use With (Application)For use when samples do not need to be transported
ColorPlain
Diameter (Metric) Tip5 mm
GradeClass II A
Length (Metric)150 mm
No. per Pack1000
Quantity1000Pack
Product TypeSwab
Unit SizePK