SG Series Gradient Makers
Actual product may vary
Fisherbrand
SG Series Gradient Makers
Ideal for generating gradients of polyacrylamide, sucrose, and cesium chloride
Have Questions?
Catalog NumberVolume (Metric)
FKS1153442415 mL
Catalog number FKS11534424
Price (SGD)
2,117.00
Each
Volume (Metric):
15 mL
Price (SGD)
2,117.00
Each
Models SG 15, SG 30, SG 50, and SG 100
  • Milled of heavy acrylic plastic with high-quality, leak-free PTFE-resin valves
  • Offer a flat base for stability on magnetic stirrers; support rod can be attached to ring stand for additional stability
  • Accept Luer or 4 mm ID tubing; with included adapters can also accept 2 mm ID or 22-gauge tubing
  • SG 100 is ideal for the Hoefer Mighty Small™ Gel Casters SE 275 (for four gels) or SE 215 (for 10 gels)
Model SG 500
  • Made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base
  • Uses a simple push-pull valve to open and close the passage between the reservoir and mixing chambers
  • Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns
  • Ideal for use with the Hoefer SE 615 or SE 915 Multiple Gel Casters
Specifications
For Use With (Equipment)Mighty Small™ Gel Caster SE 275 (for 4 Gels) or SE 215 (for 10 Gels)
TypeGradient Maker
IncludesGradient maker w/PTFE Valves , Luer outlet fitting, support rod, adapter barbed fitting and 22G needle
Volume (English)0.5 oz.
Volume (Metric)15 mL
Unit SizeEach