Small Hub Removable Needles
Small Hub Removable Needles
Actual product may vary
Small Hub Removable Needles
Available in custom lengths and point styles to fit your laboratory needs. Hamilton™ Small Hub Removable Needles are manufactured from stainless steel.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeedle GaugeNeedle Point Style
FKS1153033233 G2, 3, or 4
FKS1077606330 G2, 3, or 4
Catalog number FKS11530332
Price (SGD)
1,774.00
PK
Add to cart
Needle Gauge:
33 G
Needle Point Style:
2, 3, or 4
Price (SGD)
1,774.00
PK
Add to cart
  • Autoclavable
  • Standard needle length is 2"/51mm
  • Can be used with (RN)-style syringes that are 100μL and smaller
Specifications
MaterialStainless Steel
Needle Gauge33 G
Diameter (Metric) Inner0.11 mm
Diameter (Metric) Outer0.21 mm
Product TypeRemovable Custom Needle
Needle Point Style2, 3, or 4
For Use With (Equipment)Small Removable Syringes (100μL and smaller)
Unit SizePK