Stir Bar Box Set, Tapered with Retriever
Stir Bar Box Set, Tapered with Retriever
Actual product may vary
Fisherbrand
Stir Bar Box Set, Tapered with Retriever
Provides effective stirring for use in round-bottom flasks and containers with bases that are slightly curved or uneven.
Have Questions?
Catalog NumberLength (Metric)
FKS11527223195 mm
Catalog number FKS11527223
Price (SGD)
-
Length (Metric):
195 mm
  • Guaranteed inert and leak-free
  • Individually polished to stay cleaner for longer
  • Retriever molded in polypropylene plastic, autoclavable
  • Each set contains 20 PTFE tapered stir bars and one (1) retriever within a compartmented box. Set includes two (2) each of the following sizes (diameter x length):
4 x 10mm stir bars
5 x 15mm stir bars
7 x 20mm stir bars
8 x 25mm stir bars
8 x 30mm stir bars
8 x 35mm stir bars
8 x 40mm stir bars
8 x 50mm stir bars
8 x 60mm stir bars
  • Set also includes one (1) each of the following sizes (diameter x length): 10 x 70mm stir bar
10 x 80mm stir bar
10 x 300mm stir bar retriever
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
ColorWhite
DescriptionPTFE encapsulated Alnico V magnet core
Dimensions (L x W x H)195 x 95mm
For Use With (Equipment)Containers & Vessels / Stirrer Hot Plates
Height (Metric)30mm
Includes20 Tapered Stirrer Bars and 1 Flexible Retreiver
Width (Metric)95mm
CoatingPTFE
Length (Metric)195 mm
Quantity1/Ea.
TypePTFE Stirrer Bar and Retriever Box Set
Unit SizeEA