Check Order Status
Microcentrifuges, Micro 17/17R
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Microcentrifuges, Micro 17/17R
Adjustable 99-minute timer and quick-spin feature for short runs.
Have Questions?
Contact Us
Change view
Catalog Number
Refrigerated
FKS11526873
Yes
FKS11516873
No
2 Options
Catalog number
FKS11526873
Price (SGD)
19,197.00
EA
Add to cart
Refrigerated:
Yes
Price (SGD)
19,197.00
EA
Add to cart
Capacity: 48mL (24 x 1.5/2mL tubes)
Maximum speed/RCF: 13300rpm/17000xg
Adjustable 99 minute timer and quick-spin feature for short runs
Supplied with click seal rotor (24 place 1.5/2mL) and biocontainment lid
Specifications
Capacity
24 x 1.5/2 mL
Max. Speed
13,300 rpm
Max. RCF
17,000 x g
Description
Microcentrifuge Micro 17R, refrigerated
Refrigerated
Yes
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Safety Data Sheets
Share catalog number, name or link