PEEK Fingertip Union
PEEK Fingertip Union
Actual product may vary
Fisherbrand
PEEK Fingertip Union
Used as a connector for chromatography applications. Cytiva PEEK Fingertip Union 1/16 in. Male/M6 Female.
Have Questions?
Catalog NumberConnections
FKS115257241/16 in. Male to M6 Female
Catalog number FKS11525724
Price (SGD)
1,254.00
PK
Add to cart
Connections:
1/16 in. Male to M6 Female
Price (SGD)
1,254.00
PK
Add to cart
  • 1/16 in. Male/M6 Female Complete
  • Packsize 8 Material
  • Polyetheretherketone (PEEK) 
Specifications
Connections1/16 in. Male to M6 Female
Unit SizePK