Stainless Steel Micro Chattaway Spatulas
Stainless Steel Micro Chattaway Spatulas
Actual product may vary
Stainless Steel Micro Chattaway Spatulas
Scrapes and lifts materials from tiny bottles or flasks. Fisherbrand™ Stainless Steel Chattaway Spatulas are non-magnetic with a bright finish.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
FKS11523482100 mm
FKS11533482130 mm
FKS11543482150 mm
Catalog number FKS11523482
Price (SGD)
50.00
Pack
Add to cart
Length (Metric):
100 mm
Price (SGD)
50.00
Pack
Add to cart
  • Manufactured from Stainless Steel
  • Bent and rounded end scrapes and lifts materials from bottles and flasks
  • Fits into tight flasks and bottles with necks ≥ 4mm diameter
Specifications
Quantity5 Pk.
Product TypeChattaway Spatula
Width (Metric)4 mm
Diameter (Metric) Outer1.6 mm (Barrel)
Dimensions (L x W)100 x 4 mm
MaterialStainless Steel
Length (Metric)100 mm
Unit SizePack