Check Order Status
Stainless Steel Receiver For Test Sieves
Copy product name
Copy catalog number
Copy catalog number and product name
Copy link
Actual product may vary
Fisherbrand
Stainless Steel Receiver For Test Sieves
For the collection of materials processed through Fisherbrand™ Stainless Steel Test Sieves.
Have Questions?
Contact Us
Change view
Catalog Number
Diameter (Metric) Outer
FKS11517033
200 mm
FKS11502583
100 mm
2 Options
Catalog number
FKS11517033
Price (SGD)
89.00
EA
Estimated availability date
01-Oct-2025
Add to cart
Diameter (Metric) Outer:
200 mm
Price (SGD)
89.00
EA
Add to cart
Available for 100 and 200mm diameter sizes
Conforms to BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Specifications
Quantity
1 Each
Type
Sieve Receiver
Material
Stainless Steel
For Use With (Application)
Collection of sieved materials
Diameter (Metric) Outer
200 mm
Unit Size
EA
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
0 results displayed, search above for a specific certificate
Request a Certificate
Share catalog number, name or link