1x1 image pixel for data collection
Conductivity Standard, NIST Traceable, 10μS/cm, Reagecon™
Conductivity Standard, NIST Traceable, 10μS/cm, Reagecon™
Fisherbrand
Conductivity Standard, NIST Traceable, 10μS/cm, Reagecon™
Accredited to ISO 17025. Reagecon™ Conductivity Standard, NIST Traceable, 10μS/cm is an aqueous-based, non-hazardous low-level conductivity standard with a high degree of accuracy, low temperature variation and guaranteed stability through expiration, even after opening the bottle.
Have Questions?
Catalog NumberQuantity
FKS11516723500 mL
Catalog number FKS11516723
Price (SGD)
-
Quantity:
500 mL
Request bulk or custom format
Specifications
Shelf Life6 months
Certificate of AnalysisYes
Conductivity10μS/cm (±1% at 25°C)
Physical FormLiquid
For Use With (Equipment)Conductivity Meter
View more
This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.
  • Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
  • Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
  • Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
  • High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link