Check Order Status
PEEK Male Connector
Actual product may vary
Fisherbrand
PEEK Male Connector
Connect tubing of different sizes. Cytiva Life Sciences™ PEEK Male Connector is an important accessory for optimal chromatography.
Have Questions?
Catalog Number
Connections
FKS11515724
1/16 in. Female to M6 Male
Catalog number
FKS11515724
Price (SGD)
957.00
PK
Add to cart
Connections:
1/16 in. Female to M6 Male
Price (SGD)
957.00
PK
Add to cart
Chromatography accessory
Complete Packsize 2
Select from a variety of connections sizes and internal diamters
For use with chromatography tubing
Connectors made of Polyetheretherketone (PEEK)
Specifications
Type
Male Connector
Connections
1/16 in. Female to M6 Male
Material
PEEK