Cotton Swabs in Polypropylene Tubes
Cotton Swabs in Polypropylene Tubes
Actual product may vary
Fisher Catalogue
Cotton Swabs in Polypropylene Tubes
Use these swabs for taking samples. Fisherbrand Swabs in Polypropylene Tubes have a round, compact and shred resistant head.
Have Questions?
Catalog NumberdimTipType
FKS11512483
Catalog number FKS11512483
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Sterile dry swabs supplied in shockproof round bottom polypropylene tube
  • Sterilized by ethylene oxide
  • Produced under strict ISO 9002/EN 29002 standards
  • CE marked No.98 06 0119, Class II A
Specifications
Certifications/ComplianceISO 9002/EN 29002,CE Marked No.98 06 0119
ColorPlain
Diameter (Metric) Tip2.5 mm
GradeClass II A
Head TypeSwabs Have a Round, Compact and Shred-Resistant Head for Sample Taking
Length (Metric)150 mm
Quantity500/Pk
SterilitySterile
Handle MaterialPolystyrene
Product TypeTube Swab
Tip MaterialCotton
Unit SizePK