Cardboard Cryoboxes, 136mm
Cardboard Cryoboxes, 136mm
Actual product may vary
Fisherbrand
Cardboard Cryoboxes, 136mm
Combine these boxes with different grid inserts to hold a wide range of tube sizes. Fisherbrand™ Cardboard Cryoboxes are temperature resistant down to -80°C, ideal for ultra-low temperature storage.
Have Questions?
Catalog NumberDimensions (L x W x H)Color
FKS11507633136 x 136 x 75 mmWhite
Catalog number FKS11507633
Price (SGD)
139.00
PK
Add to cart
Dimensions (L x W x H):
136 x 136 x 75 mm
Color:
White
Price (SGD)
139.00
PK
Add to cart
  • Available with a water-repellent finish or with a waterproof plastic coating
  • Highest flexibility: Cryoboxes can be combined with different grid inserts for tubes from 6 to 28.5mm in diameter
  • The Cryoboxes can be used for larger vessels if the grid is removed
Specifications
Capacity (Metric)4 mL
ColorWhite
MaterialCardboard
ShapeSquare
TypeCryo Box
Quantity10 Pack
Dimensions (L x W)136 x 136 mm
Dimensions (L x W x H)136 x 136 x 75 mm
Length (Metric)136 mm
Width (Metric)136 mm
Width (Metric) Interior136 mm
Height (Metric)75 mm
Weight (Metric)1.1 kg
Unit SizePK