Stainless Steel Chattaway Spatulas
Stainless Steel Chattaway Spatulas
Actual product may vary
Fisher Catalogue
Stainless Steel Chattaway Spatulas
Designed using high quality stainless steel and nickel components these trough spatulas are ideal in most laboratory, research, and other environments.
Have Questions?
Catalog NumberLength (Metric)
FKS11503492150 mm
Catalog number FKS11503492
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
150 mm
  • Spatulas, nickel and stainless steel Chattaway 3.6mm barrel, nickel and stainless micros 1.6mm barrel.
  • Trough spatula, trough bladed end measuring 8mm wide x 50mm long x 1.5mm deep
  • Manufactured from 304 spec stainless steel
  • For sterile applications
  • Uses the finest in corrosion-resistant 304 stainless steel
  • Available in standard packs of 5
Specifications
Quantity5 Pk.
SterilitySterile
AutoclavableAutoclavable
Product TypeTrough Spatula
For Use With (Application)For Sterile Applications
Width (Metric) Blade8 mm
Temperature (Metric) Max-196°C to +550°C
MaterialStainless Steel
Length (Metric)150 mm
Unit SizeEA