Tube Closures
Actual product may vary
Fisherbrand
Tube Closures
Suitable for different usage of tubes and applications. Gosselin™ Tube Closures are offered in straight, winged, skirted, or screw cap styles and are manufactured from PE plastic.
Have Questions?
Catalog NumberColorProduct TypeFor Use With (Equipment)Quantity
FKS11502772WhiteStraight Plug13.5 mm Tube
Catalog number FKS11502772
Price (SGD)
421.00
Pack
Color:
White
Product Type:
Straight Plug
For Use With (Equipment):
13.5 mm Tube
Price (SGD)
421.00
Pack
  • All closures vertically ribbed for easier grip
  • Protected with a polyethylene outer bag to prevent particle contamination
Specifications
Certifications/ComplianceEC N°1935/2004, ISO 11137
Product TypeStraight Plug
MaterialLDPE
ColorWhite
For Use With (Equipment)13.5 mm Tube
Unit SizePack