Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf
Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf
Actual product may vary
Fisher Catalogue
Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf
Get maximum observation of samples, water circulation, and temperature stability with the stirred water bath. Designed for immersion of flasks, bottles, racks in a highly stable temperature environment, this Clifton™ Digital Thermostirrer with Stainless Steel Tank and Shelf includes anti-bacterial technology and other unique features as demanded by many clinical and laboratory environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS1149950922 L
FKS1148950914 L
Catalog number FKS11499509
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
22 L
  • Stirred Clifton water bath designed for immersion of flasks, bottles, racks in a highly stable temperature environment
  • Thermostirrer is robust - constructed from stainless steel for durability
  • Digital PID temperature control
  • Easy to use controls that wipe clean and have an anti-bacterial finish
  • Sturdy clear co-polyester BPA free tank allows observation of samples
  • Thermostirrer controls and painted surfaces feature “anti-bacterial technology” 
  • When low liquid level is detected, a “FiLL” message automatically appears and heating is suspended
  • Every unit undergoes a factory 2 point calibration procedure 
Specifications
DescriptionDigital Water Bath
Capacity (Metric)22 L
Certifications/ComplianceCE Approved
Temperature Range5°C to 99°C
DisplayDigital LED
MaterialStainless Steel
ModelNE4-22 D
Temperature Resolution±0.1°C
Warranty3 Years
Dimensions (D x W x H)332 x 537 x 170 mm
Dimensions (D x W x H) Interior298 x 395 x 150 mm
Width (Metric)537 mm
Width (Metric) Interior395 mm
Height (Metric)170 mm
Height (Metric) Interior150 mm
Depth (Metric)332 mm
Depth (Metric) Interior298 mm
Weight (Metric)12.7 kg
Voltage230 V
Wattage1250 W