Interchangeable Block for QB Series Dry Block Heaters
Interchangeable Block for QB Series Dry Block Heaters
Actual product may vary
Fisher Catalogue
Interchangeable Block for QB Series Dry Block Heaters
For use with 0.2mL tubes, strips and 96-well microtitre plates used in molecular biology and biotechnology applications. Grant Instruments™ Interchangeable Block for QB Series Dry Block Heaters can withstand temperatures up to 100°C.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTube SizeNo. of PlacesDepth (Metric)
FKS114909181.5mL2435 mm hole depth
FKS114009282.0mL2435 mm hole depth
Catalog number FKS11490918
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Tube Size:
1.5mL
No. of Places:
24
Depth (Metric):
35 mm hole depth
  • For use with QBD1, QBD2, QBD4, QBH2, QBA1 and QBA2 dry block heating system
  • Accommodates a variety of test tube sizes
  • Available in plain block format without holes
Specifications
For Use With (Equipment)10.8mm dia. microtubes
MaterialAluminum
No. of Places24
TypeBlock
Tube Size1.5mL
Dimensions (L x W x H)140 x 50 x 63 mm
Depth (Metric)35 mm hole depth
Unit SizeEA