Cotton Wool, Non-absorbent
Cotton Wool, Non-absorbent
Actual product may vary
Fisherbrand
Cotton Wool, Non-absorbent
Test absorbency by placing a teaspoon of water in the hollow of a cotton. Azpack™ Cotton Wool, Non-absorbent is defatted due to bleaching.
Have Questions?
Catalog NumberWeight (Metric)
FKS11468153500 g
Catalog number FKS11468153
Price (SGD)
163.00
EA
Add to cart
Weight (Metric):
500 g
Price (SGD)
163.00
EA
Add to cart
  • 100% Bleached cotton combers
  • Treated with a non-absorbent finish
  • 1 roll is approximately 3.5m x 325mm (L x W)
  • 500g
  • CE Marked
Specifications
Product TypeNonabsorbent Cotton
MaterialBleached Cotton treated with nonabsorbent finish
GradeCE Marked
Quantity1 Ea.
Weight (Metric)500 g
Unit SizeEA