Test Tube Racks
Test Tube Racks
Actual product may vary
Fisherbrand
Test Tube Racks
Customize your test tube rack to fit a varietry of test tube external diameters. Nickel Electro™ Test Tube Racks are non-magnetic stainless steel and supplied flat packed ready for assembly.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHolds
FKS11449529Tubes (31 mm)
FKS11429529Tubes (18 mm)
Catalog number FKS11449529
Price (SGD)
323.00
EA
Add to cart
Holds:
Tubes (31 mm)
Price (SGD)
323.00
EA
Add to cart
  • Stainless steel
  • Non-magnetic
  • Easy assembley
  • Capable of being disassembled for cleaning and sterilization purposes
Specifications
Product TypeTest Tube Rack
AutoclavableAutoclavable
HoldsTubes (31 mm)
MaterialStainless Steel
Positions12
For Use With (Equipment)Test Tubes with an External Diameter 31 mm
Quantity1 Ea.
Dimensions (L x W x H)270 x 138 x 70 mm
Diameter (Metric) Opening31 mm
Diameter (Metric) Well32 mm
Unit SizeEA