Stuart™  Hot Plate, Ambient to 300°C, Silumin
Actual product may vary
Fisherbrand
Stuart™ Hot Plate, Ambient to 300°C, Silumin
Shape: Rectangle; Size: 11.8 x 19.7 in.
Have Questions?
Catalog NumberController TypeHeight (Metric)
FKS11446518Analog130 mm
Catalog number FKS11446518
Price (SGD)
-
Controller Type:
Analog
Height (Metric):
130 mm
Shape: Rectangle; Size: 11.8 x 19.7 in.
  • Durable alloy plate top.
  • Even and consistent heating.
  • Temperature controlled with an easy to use dial.
  • Fitted with a HOT warning light for temperatures over 50°C.
Specifications
Top Plate MaterialSilumin
Controller TypeAnalog
Size11.8 x 19.7 in.
Temperature (Metric)Ambient to 300°C
Depth (Metric)360 mm
Height (Metric)130 mm
Weight (Metric)12 kg
Width (Metric)520 mm
Wattage1500 w
Electrical Requirements230 V, 50/60 Hz, 1500 w
Frequency50/60 Hz
ModelSB500
ShapeRectangle
Voltage230 V
TypeHot Plate
DisplayAnalog
Dimensions (L x W x H)11.8 in. x 19.7 in.
Unit SizeEach